Ethyl(3,4-dimethoxybenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3,4-dimethoxyphenyl)-2-oxoacetate

023021-1G
FLUO023021-1GEA 0 EUR
FLUO023021-1G
Ethyl(3,4-dimethoxybenzoyl)formiat
Ethyl(3,4-dimethoxybenzoyl)formiat
Formel: C₁₂H₁₄O₅
CAS-Nummer: 40233-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR