Ethyl(4-isopropylbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-isopropylphenyl)-2-oxoacetate

023022-1G
FLUO023022-1GEA 104 EUR
FLUO023022-1G
Ethyl(4-isopropylbenzoyl)formiat
Ethyl(4-isopropylbenzoyl)formiat
Formel: C₁₃H₁₆O₃
Molecular Weight: 220,2643 g/mol
MDL: MFCD06201711
CAS-Nummer: 34906-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR