Ethyl(4-chlor-3-fluorbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-chloro-3-fluorophenyl)-2-oxoacetate

023023-1G
FLUO023023-1GEA 882 EUR
FLUO023023-1G
Ethyl(4-chlor-3-fluorbenzoyl)formiat
Ethyl(4-chlor-3-fluorbenzoyl)formiat
Formel: C₁₀H₈ClFO₃
Molecular Weight: 230,62 g/mol
MDL: MFCD06201712
CAS-Nummer: 845790-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR