Ethyl(4-fluor-3-methylbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-fluoro-3-methylphenyl)-2-oxoacetate

023024-1G
FLUO023024-1GEA 647 EUR
FLUO023024-1G
Ethyl(4-fluor-3-methylbenzoyl)formiat
Ethyl(4-fluor-3-methylbenzoyl)formiat
Formel: C₁₁H₁₁FO₃
CAS-Nummer: 732251-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR