Ethyl(4-(dimethylamino)benzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-(dimethylamino)phenyl)-2-oxoacetate

023025-1G
FLUO023025-1GEA 90.9 EUR
FLUO023025-1G
Ethyl(4-(dimethylamino)benzoyl)formiat
Ethyl(4-(dimethylamino)benzoyl)formiat
Formel: C₁₂H₁₅NO₃
CAS-Nummer: 41116-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR