Ethyl(4-ethylbenzoyl)formiat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-(4-ethylphenyl)-2-oxoacetate
023026-1G
FLUO023026-1GEA
647
EUR
FLUO023026-1G
Ethyl(4-ethylbenzoyl)formiat
Ethyl(4-ethylbenzoyl)formiat
Formel:
C₁₂H₁₄O₃ Molecular Weight: 206.238 g/mol |
MDL:
MFCD06201713 CAS-Nummer: 62936-36-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...