Ethyl(3-chlor-5-fluorbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(3-chloro-5-fluorophenyl)-2-oxoacetate

023027-1G
FLUO023027-1GEA 527 EUR
FLUO023027-1G
Ethyl(3-chlor-5-fluorbenzoyl)formiat
Ethyl(3-chlor-5-fluorbenzoyl)formiat
Formel: C₁₀H₈ClFO₃
Molecular Weight: 230,62 g/mol
MDL: MFCD06201714
CAS-Nummer: 845790-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR