Ethyl(3-methylbenzoyl)formiat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-oxo-2-(m-tolyl)acetate
023028-1G
FLUO023028-1GEA
0
EUR
FLUO023028-1G
Ethyl(3-methylbenzoyl)formiat
Ethyl(3-methylbenzoyl)formiat
Formel:
C₁₁H₁₂O₃ Molecular Weight: 192,211 g/mol |
MDL:
MFCD01311168 CAS-Nummer: 66644-68-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...