Ethyl(4-chlorbenzoyl)formiat

Lieferant: FLUOROCHEM

023029-1G
FLUO023029-1GEA 31.1 EUR
FLUO023029-1G
Ethyl(4-chlorbenzoyl)formiat
Ethyl(4-chlorbenzoyl)formiat
Formel: C₁₀H₉ClO₃
MDL: MFCD01319615
CAS-Nummer: 34966-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR