Ethyl(4-(methylthio)benzoyl)formiat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 2-(4-(methylthio)phenyl)-2-oxoacetate
023030-1G
FLUO023030-1GEA
279
EUR
FLUO023030-1G
Ethyl(4-(methylthio)benzoyl)formiat
Ethyl(4-(methylthio)benzoyl)formiat
Formel:
C₁₁H₁₂O₃S Molecular Weight: 224,276 g/mol |
MDL:
MFCD01319625 CAS-Nummer: 62936-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...