Ethyl(4-chlor-3-methylbenzoyl)formiat

Lieferant: FLUOROCHEM

Synonyme: Ethyl 2-(4-chloro-3-methylphenyl)-2-oxoacetate

023032-1G
FLUO023032-1GEA 0 EUR
FLUO023032-1G
Ethyl(4-chlor-3-methylbenzoyl)formiat
Ethyl(4-chlor-3-methylbenzoyl)formiat
Formel: C₁₁H₁₁ClO₃
Molecular Weight: 226,656 g/mol
MDL: MFCD06201715
CAS-Nummer: 250642-57-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR