4,4',4''-Trimethyltritylalkohol

Lieferant: FLUOROCHEM

Synonyme: Tri-p-tolylmethanol

023044-1G
FLUO023044-1GEA 0 EUR
FLUO023044-1G
4,4',4''-Trimethyltritylalkohol
4,4',4''-Trimethyltritylalkohol
Formel: C₂₂H₂₂O
Molecular Weight: 302,42 g/mol
MDL: MFCD00014919
CAS-Nummer: 3247-00-5
EINECS: 221-822-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR