3-Fluor-4-methoxythiophenol

Lieferant: FLUOROCHEM

023073-1G
FLUO023073-1GEA 956 EUR
FLUO023073-1G
3-Fluor-4-methoxythiophenol
3-Fluor-4-methoxythiophenol
Formel: C₇H₇FOS
Molecular Weight: 158.1933 g/mol
MDL: MFCD06201751
CAS-Nummer: 89818-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR