3,4-(Ethylendioxy)thiophenol
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dihydrobenzo[b][1,4]dioxine-6-thiol
023075-1G
FLUO023075-1GEA
981
EUR
FLUO023075-1G
3,4-(Ethylendioxy)thiophenol
3,4-(Ethylendioxy)thiophenol
Formel:
C₈H₈O₂S Molecular Weight: 168,212 g/mol |
MDL:
MFCD06201753 CAS-Nummer: 247228-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...