3,4-(Methylendioxy)thiophenol

Lieferant: FLUOROCHEM

023078-1G
FLUO023078-1GEA 1090 EUR
FLUO023078-1G
3,4-(Methylendioxy)thiophenol
3,4-(Methylendioxy)thiophenol
Formel: C₇H₆O₂S
MDL: MFCD06201755
CAS-Nummer: 5274-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR