4-Fluor-m-kresol

Lieferant: FLUOROCHEM
Achtung

023080-1G
FLUO023080-1GEA 290 EUR
FLUO023080-1G
4-Fluor-m-kresol
4-Fluor-m-kresol
Formel: C₇H₇FO
MDL: MFCD00042296
CAS-Nummer: 452-70-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR