3-(4-Methylphenyl)thiophenol
Lieferant: FLUOROCHEM
023097-1G
FLUO023097-1GEA
1200
EUR
FLUO023097-1G
3-(4-Methylphenyl)thiophenol
3-(4-Methylphenyl)thiophenol
Formel:
C₁₃H₁₂S |
MDL:
MFCD06201774 CAS-Nummer: 933674-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...