3,5-Difluorthiophenol
Lieferant: FLUOROCHEM
Synonyme:
3,5-Difluorbenzolthiol, 3,5-Difluorphenylmercaptan, 1,3-Difluor-5-mercaptobenzol, 3,5-Difluorbenzol-1-thiol
023121-1G
FLUO023121-1GEA
31.1
EUR
FLUO023121-1G
3,5-Difluorthiophenol
3,5-Difluorthiophenol
Formel:
C₆H₄F₂S Molecular Weight: 146,16 g/mol |
MDL:
MFCD06201795 CAS-Nummer: 99389-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...