3-Chlor-5-fluorthiophenol

Lieferant: FLUOROCHEM

023127-1G
FLUO023127-1GEA 432 EUR
FLUO023127-1G
3-Chlor-5-fluorthiophenol
3-Chlor-5-fluorthiophenol
Formel: C₆H₄ClFS
MDL: MFCD06201798
CAS-Nummer: 845823-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR