4-Fluor-2-methylthiophenol

Lieferant: FLUOROCHEM

Synonyme: 4-Fluoro-2-methylthiophenol, 4-Fluoro-2-(methylthio)phenol

023130-1G
FLUO023130-1GEA 554 EUR
FLUO023130-1G
4-Fluor-2-methylthiophenol
4-Fluor-2-methylthiophenol
Formel: C₇H₇FOS
Molecular Weight: 158.19 g/mol
MDL: MFCD16998914
CAS-Nummer: 90033-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR