4'-tert-Butyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-(Tert-butyl)phenyl)-2,2,2-trifluoroethanone

023134-1G
FLUO023134-1GEA 209 EUR
FLUO023134-1G
4'-tert-Butyl-2,2,2-trifluoracetophenon
4'-tert-Butyl-2,2,2-trifluoracetophenon
Formel: C₁₂H₁₃F₃O
MDL: MFCD01319979
CAS-Nummer: 73471-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR