4'-Methyl-2,2,2,3'-tetrafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(3-fluoro-4-methylphenyl)ethanone

023136-1G
FLUO023136-1GEA 222 EUR
FLUO023136-1G
4'-Methyl-2,2,2,3'-tetrafluoracetophenon
4'-Methyl-2,2,2,3'-tetrafluoracetophenon
Formel: C₉H₆F₄O
MDL: MFCD01320003
CAS-Nummer: 845823-06-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR