4'-Methoxy-2,2,2,3'-tetrafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(3-fluoro-4-methoxyphenyl)ethanone

023137-1G
FLUO023137-1GEA 341 EUR
FLUO023137-1G
4'-Methoxy-2,2,2,3'-tetrafluoracetophenon
4'-Methoxy-2,2,2,3'-tetrafluoracetophenon
Formel: C₉H₆F₄O₂
MDL: MFCD06201802
CAS-Nummer: 132734-54-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR