4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV )

Lieferant: FLUOROCHEM

Synonyme: 4'-Butyl-2,2,2-trifluoroacetophenone, 1-(4-Butylphenyl)-2,2,2-trifluoroethanone

023140-1G
FLUO023140-1GEA 594 EUR
FLUO023140-1G
4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV )
4'-Butyl-2,2,2-trifluoracetophenon (Carbonat Ionophor IV )
Formel: C₁₂H₁₃F₃O
MDL: MFCD00214157
CAS-Nummer: 40739-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR