3'-Chlor-2,2,2,5'-tetrafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Chloro-5-fluorophenyl)-2,2,2-trifluoroethanone

023141-1G
FLUO023141-1GEA 0 EUR
FLUO023141-1G
3'-Chlor-2,2,2,5'-tetrafluoracetophenon
3'-Chlor-2,2,2,5'-tetrafluoracetophenon
Formel: C₈H₃ClF₄O
MDL: MFCD02260831
CAS-Nummer: 845823-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR