4'-(Ethylthio)-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-(ethylthio)phenyl)-2,2,2-trifluoroethanone

023146-1G
FLUO023146-1GEA 227 EUR
FLUO023146-1G
4'-(Ethylthio)-2,2,2-trifluoracetophenon
4'-(Ethylthio)-2,2,2-trifluoracetophenon
Formel: C₁₀H₉F₃OS
Molecular Weight: 234.238 g/mol
MDL: MFCD06201804
CAS-Nummer: 845823-09-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR