3'-Methoxy-2,2,2-trifluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
2,2,2-Trifluoro-1-(3-methoxyphenyl)ethanone
023148-1G
FLUO023148-1GEA
159
EUR
FLUO023148-1G
3'-Methoxy-2,2,2-trifluoracetophenon
3'-Methoxy-2,2,2-trifluoracetophenon
Formel:
C₉H₇F₃O₂ Molecular Weight: 204,1459 g/mol |
MDL:
MFCD00461895 CAS-Nummer: 30724-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...