3'-Methyl-2,2,2,4'-tetrafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(4-fluoro-3-methylphenyl)ethanone

023149-1G
FLUO023149-1GEA 279 EUR
FLUO023149-1G
3'-Methyl-2,2,2,4'-tetrafluoracetophenon
3'-Methyl-2,2,2,4'-tetrafluoracetophenon
Formel: C₉H₆F₄O
MDL: MFCD01320004
CAS-Nummer: 845823-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR