4'-Methoxy-2'-methyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

023150-1G
FLUO023150-1GEA 558 EUR
FLUO023150-1G
4'-Methoxy-2'-methyl-2,2,2-trifluoracetophenon
4'-Methoxy-2'-methyl-2,2,2-trifluoracetophenon
Formel: C₁₀H₉F₃O₂
MDL: MFCD03844198
CAS-Nummer: 845823-11-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR