3'-Isopropyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(3-isopropylphenyl)ethanone

023153-1G
FLUO023153-1GEA 177 EUR
FLUO023153-1G
3'-Isopropyl-2,2,2-trifluoracetophenon
3'-Isopropyl-2,2,2-trifluoracetophenon
Formel: C₁₁H₁₁F₃O
MDL: MFCD06201806
CAS-Nummer: 155628-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR