3'-Chlor-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3-Chlorophenyl)-2,2,2-trifluoroethanone

023154-1G
FLUO023154-1GEA 101 EUR
FLUO023154-1G
3'-Chlor-2,2,2-trifluoracetophenon
3'-Chlor-2,2,2-trifluoracetophenon
Formel: C₈H₄ClF₃O
Molecular Weight: 208,5650 g/mol
MDL: MFCD00461896
CAS-Nummer: 321-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR