2,2,2,3',5'-Pentafluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Difluorophenyl)-2,2,2-trifluoroethanone

023155-1G
FLUO023155-1GEA 34.1 EUR
FLUO023155-1G
2,2,2,3',5'-Pentafluoracetophenon
2,2,2,3',5'-Pentafluoracetophenon
Formel: C₈H₃F₅O
Molecular Weight: 210,1000 g/mol
MDL: MFCD01319993
CAS-Nummer: 845823-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR