3',5'-Dichlor-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Dichlorophenyl)-2,2,2-trifluoroethanone

023161-1G
FLUO023161-1GEA 61.2 EUR
FLUO023161-1G
3',5'-Dichlor-2,2,2-trifluoracetophenon
3',5'-Dichlor-2,2,2-trifluoracetophenon
Formel: C₈H₃Cl₂F₃O
Molecular Weight: 243,0100 g/mol
MDL: MFCD01319996
CAS-Nummer: 130336-16-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR