4'-Chlor-2'-methyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chloro-2-methylphenyl)-2,2,2-trifluoroethanone

023162-1G
FLUO023162-1GEA 290 EUR
FLUO023162-1G
4'-Chlor-2'-methyl-2,2,2-trifluoracetophenon
4'-Chlor-2'-methyl-2,2,2-trifluoracetophenon
Formel: C₉H₆ClF₃O
Molecular Weight: 222,60 g/mol
MDL: MFCD06201807
CAS-Nummer: 845823-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR