4'-Chlor-2,2,2,3'-tetrafluoracetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(4-Chloro-3-fluorophenyl)-2,2,2-trifluoroethanone
023164-1G
FLUO023164-1GEA
519
EUR
FLUO023164-1G
4'-Chlor-2,2,2,3'-tetrafluoracetophenon
4'-Chlor-2,2,2,3'-tetrafluoracetophenon
Formel:
C₈H₃ClF₄O Molecular Weight: 226,55 g/mol |
MDL:
MFCD01319997 CAS-Nummer: 845823-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...