4'-Isopropyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-1-(4-isopropylphenyl)ethanone

023166-1G
FLUO023166-1GEA 29.3 EUR
FLUO023166-1G
4'-Isopropyl-2,2,2-trifluoracetophenon
4'-Isopropyl-2,2,2-trifluoracetophenon
Formel: C₁₁H₁₁F₃O
MDL: MFCD01319978
CAS-Nummer: 124211-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR