4'-Chlor-3'-methyl-2,2,2-trifluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-Chloro-3-methylphenyl)-2,2,2-trifluoroethanone

023167-1G
FLUO023167-1GEA 648 EUR
FLUO023167-1G
4'-Chlor-3'-methyl-2,2,2-trifluoracetophenon
4'-Chlor-3'-methyl-2,2,2-trifluoracetophenon
Formel: C₉H₆ClF₃O
MDL: MFCD02260828
CAS-Nummer: 286017-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR