3-(Trimethylsilyl)anisol

Lieferant: FLUOROCHEM

023173-1G
FLUO023173-1GEA 327 EUR
FLUO023173-1G
3-(Trimethylsilyl)anisol
3-(Trimethylsilyl)anisol
Formel: C₁₀H₁₆OSi
MDL: MFCD06201812
CAS-Nummer: 17876-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR