4-(Trimethylsilyl)anisol

Lieferant: FLUOROCHEM

Synonyme: (4-Methoxyphenyl)trimethylsilane

023175-1G
FLUO023175-1GEA 689 EUR
FLUO023175-1G
4-(Trimethylsilyl)anisol
4-(Trimethylsilyl)anisol
Formel: C₁₀H₁₆OSi
Molecular Weight: 180,319 g/mol
MDL: MFCD06201814
CAS-Nummer: 877-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR