2-Brom-1-dodecen
Lieferant: FLUOROCHEM
023214-1G
FLUO023214-1GEA
942
EUR
FLUO023214-1G
2-Brom-1-dodecen
2-Bromo-1-dodecene
Formel:
C₁₂H₂₃Br |
CAS-Nummer:
74571-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...