4'-(2-Fluorphenoxy)acetophenon

Lieferant: FLUOROCHEM

023219-25G
FLUO023219-25GEA 684 EUR
FLUO023219-25G
4'-(2-Fluorphenoxy)acetophenon
4'-(2-Fluorphenoxy)acetophenon
Formel: C₁₄H₁₁FO₂
MDL: MFCD06411644
CAS-Nummer: 58775-91-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR