2'-(3-Fluorbenzyloxy)acetophenon

Lieferant: FLUOROCHEM

023221-25G
FLUO023221-25GEA 942 EUR
FLUO023221-25G
2'-(3-Fluorbenzyloxy)acetophenon
2'-(3-Fluorbenzyloxy)acetophenon
Formel: C₁₅H₁₃FO₂
MDL: MFCD06411645
CAS-Nummer: 845823-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR