(±)-2-Methylpiperazin

Lieferant: FLUOROCHEM

Synonyme: 2-Methylpiperazine

023229-100G
FLUO023229-100GEA 0 EUR
FLUO023229-100G
(±)-2-Methylpiperazin
(±)-2-Methylpiperazin
Formel: C₅H₁₂N₂
Molecular Weight: 100,16 g/mol
Siedepunkt: 155 °C (1013 hPa)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005954
CAS-Nummer: 109-07-9
EINECS: 203-644-2
UN: 2925
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR