Mucobromsäure
Lieferant: FLUOROCHEM
Synonyme:
2,3-Dibromomalealdehydic acid, (Z)-2,3-Dibromo-4-oxobut-2-enoic acid
023257-100G
023257-25G
FLUO023257-100GEA
81.9
EUR
FLUO023257-100G
FLUO023257-25G
Mucobromsäure
Mucobromsäure
Formel:
C₄H₂Br₂O₃ Molecular Weight: 257.88 g/mol |
MDL:
MFCD00063745 CAS-Nummer: 488-11-9 EINECS: 207-670-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...