Mucobromsäure

Lieferant: FLUOROCHEM

Synonyme: 2,3-Dibromomalealdehydic acid, (Z)-2,3-Dibromo-4-oxobut-2-enoic acid

023257-100G 023257-25G
FLUO023257-100GEA 81.9 EUR
FLUO023257-100G FLUO023257-25G
Mucobromsäure
Mucobromsäure
Formel: C₄H₂Br₂O₃
Molecular Weight: 257.88 g/mol
MDL: MFCD00063745
CAS-Nummer: 488-11-9
EINECS: 207-670-5
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR