Benzyl-phenylether

Lieferant: FLUOROCHEM

Synonyme: Benzyl Phenyl Ether

023290-25G
FLUO023290-25GEA 103 EUR
FLUO023290-25G
Benzyl-phenylether
Benzyl-phenylether
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
MDL: MFCD00020660
CAS-Nummer: 946-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR