2-Amino-3-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

023329-1G
FLUO023329-1GEA 31.1 EUR
FLUO023329-1G
2-Amino-3-(trifluormethyl)pyridin
2-Amino-3-(trifluoromethyl)pyridine
Formel: C₆H₅F₃N₂
Molecular Weight: 162,11 g/mol
MDL: MFCD04972538
CAS-Nummer: 183610-70-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR