3-Methylbenzo[b]thiophen-2-carbaldehyd

Lieferant: FLUOROCHEM

023335.5G 023335-1G
FLUO023335.5GEA 585 EUR
FLUO023335.5G FLUO023335-1G
3-Methylbenzo[b]thiophen-2-carbaldehyd
3-Methylbenzo[b]thiophen-2-carbaldehyd
Formel: C₁₀H₈OS
Molecular Weight: 176,24 g/mol
MDL: MFCD00052307
CAS-Nummer: 22053-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR