4-Ethylphenylhydrazin Hydrochlorid

Lieferant: FLUOROCHEM

Synonyme: 4-Ethylphenylhydrazine Hydrochloride

023498-1G
FLUO023498-1GEA 39.8 EUR
FLUO023498-1G
4-Ethylphenylhydrazin Hydrochlorid
4-Ethylphenylhydrazin Hydrochlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172.6552 g/mol
MDL: MFCD01861267
CAS-Nummer: 53661-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR