4-Chloro-2-fluorophenylhydrazine hydrochloride

Lieferant: FLUOROCHEM

Synonyme: (4-Chloro-2-fluorophenyl)hydrazine hydrochloride

023512-1G
FLUO023512-1GEA 31.1 EUR
FLUO023512-1G
4-Chloro-2-fluorophenylhydrazine hydrochloride
4-Chloro-2-fluorophenylhydrazine hydrochloride
Formel: C₆H₆ClFN₂·ClH
Molecular Weight: 197,038 g/mol
MDL: MFCD06245512
CAS-Nummer: 64172-78-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR